• USA Home
  • B6652 - 1-(Boc-amino)cyclopropanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
B6652 Sigma-Aldrich

1-(Boc-amino)cyclopropanecarboxylic acid

≥98%

  • CAS Number 88950-64-5

  • Empirical Formula (Hill Notation) C9H15NO4

  • Molecular Weight 201.22

  •  Beilstein/REAXYS Number 4430628

  •  MDL number MFCD00083257

  •  PubChem Substance ID 24891927

Purchase

Properties

assay   ≥98%
storage temp.   2-8°C
SMILES string   CC(C)(C)OC(=O)NC1(CC1)C(O)=O
InChI   1S/C9H15NO4/c1-8(2,3)14-7(13)10-9(4-5-9)6(11)12/h4-5H2,1-3H3,(H,10,13)(H,11,12)
InChI key   DSKCOVBHIFAJRI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?