EMAIL THIS PAGE TO A FRIEND
B6881 Sigma-Aldrich

Biuret

≥98%

Synonym: Allophanic acid amide, Carbamoylurea

  • CAS Number 108-19-0

  • Linear Formula NH2CONHCONH2

  • Molecular Weight 103.08

  •  Beilstein/REAXYS Number 1703510

  •  EC Number 203-559-0

  •  MDL number MFCD00007946

Purchase

Properties

assay   ≥98%
mp   185-190 °C (dec.) (lit.)
SMILES string   NC(=O)NC(N)=O
InChI   1S/C2H5N3O2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)
InChI key   OHJMTUPIZMNBFR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?