EMAIL THIS PAGE TO A FRIEND
B7006 Sigma-Aldrich

Boc-Cys(Trt)-OH

Synonym: N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, Nα-Boc-S-trityl-L-cysteine

  • CAS Number 21947-98-8

  • Linear Formula (C6H5)3CSCH2CH[NHCO2C(CH3)3]CO2H

  • Molecular Weight 463.59

  •  Beilstein/REAXYS Number 2028965

  •  EC Number 244-674-6

  •  MDL number MFCD00038251

Purchase

Properties

mp   143-146 °C (lit.)
storage temp.   −20°C
SMILES string   CC(C)(C)OC(=O)N[C@@H](CSC(c1ccccc1)(c2ccccc2)c3ccccc3)C(O)=O
InChI   1S/C27H29NO4S/c1-26(2,3)32-25(31)28-23(24(29)30)19-33-27(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18,23H,19H2,1-3H3,(H,28,31)(H,29,30)/t23-/m0/s1
InChI key   JDTOWOURWBDELG-QHCPKHFHSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?