EMAIL THIS PAGE TO A FRIEND
B8548 Sigma-Aldrich

Bromine

BioXtra, 99.5-101.0%

Purchase

Properties

vapor density   7.14 (vs air)
vapor pressure   175 mmHg ( 20 °C)
  671 mmHg ( 55 °C)
product line   BioXtra
assay   99.5-101.0%
resistivity   7.8E18 μΩ-cm, 20°C
impurities   <0.0005% Phosphorus (P)
  <0.001% Sulfur compounds
evapn. residue   <0.005%
bp   58.8 °C (lit.)
mp   −7.2 °C (lit.)
density   3.119 g/mL at 25 °C (lit.)
anion traces   chloride (Cl-): <0.05%
cation traces   Al: <0.0005%
  Ca: <0.0005%
  Cu: <0.0005%
  Fe: <0.005%
  K: <0.005%
  Mg: <0.0005%
  Na: <0.005%
  Pb: <0.001%
  Zn: <0.0005%
storage temp.   2-8°C
SMILES string   BrBr
InChI   1S/Br2/c1-2
InChI key   GDTBXPJZTBHREO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1744 8(6.1) / PGI
WGK Germany 
2
RTECS 
EF9100000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

470864 Bromine, ≥99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?