EMAIL THIS PAGE TO A FRIEND
B9518 Sigma-Aldrich

BigCHAP

~98%

Synonym: N,N-Bis[3-(D-gluconamido)propyl]cholamide

  • CAS Number 86303-22-2

  • Empirical Formula (Hill Notation) C42H75N3O16

  • Molecular Weight 878.06

  •  MDL number MFCD00082541

Purchase

Properties

description   non-ionic
assay   ~98%
storage temp.   2-8°C

Description

Application

Useful in solubilization of functional opiate receptor.

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?