EMAIL THIS PAGE TO A FRIEND
B9774 Sigma

Boc-His(Dnp)-OH

Synonym: Nα-Boc-N(im)-2,4-dinitrophenyl-L-histidine

  • CAS Number 25024-53-7

  • Empirical Formula (Hill Notation) C17H19N5O8

  • Molecular Weight 421.36

  •  Beilstein Registry Number 771922

  •  EC Number 246-569-0

  •  MDL number MFCD00065966

Purchase

Properties

InChI Key   KPGVUOQMOHGHEW-LBPRGKRZSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15287 Boc-His(Dnp)-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?