• USA Home
  • BAH369040005 - Bel-Art® Sterileware Sense-able Scoops®
EMAIL THIS PAGE TO A FRIEND
BAH369040005 Sigma
Bel-Art® Sterileware Sense-able Scoops®
Metal Detectable, capacity 4 oz
Purchase
Properties
Related Categories Bel-Art, Bel-Art Spatulas, Labware, Manufacturer Browser, Scienceware polypropylene scoops,
material   (polystyrene impregnated with stainless steel powder)
  black
packaging   package of 100
mfr. no.   Bel-Art, H36904-0005
L × W × H   91 mm × 46 mm × 31 mm
capacity   4 oz
Description
General description
Why risk your company reputation and suffer costly product recalls? Plastic impregnated with stainless steel powder permits fragments to be detected with standard metal detection and X-ray equipment.
Features and Benefits
Stainless steel powder homogeneously incorporated into scoops from top to bottom
Sterilized by Gamma Irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
Long handle increases the distance between the hand and sample to reduce contamination and exposure potential
Flat bottom allows scoops to remain upright for use as weigh boats or to rest on a flat surface without spilling
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sense-able Scoops is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?