• USA Home
  • BM0025 - BMS-538305 L-(+)-tartrate

EMAIL THIS PAGE TO A FRIEND
BM0025 Sigma-Aldrich

BMS-538305 L-(+)-tartrate

≥98% (HPLC)

Synonym: (1S,5R)-2-[(2S)-amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-Azabicyclo[3.1.0]hexane L-(+)-Tartaric acid salt, (2S)-2-Amino-1-(1S,5R)-2-azabicyclo[3.1.0]hex-2-yl-2-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-ethanone L-(+)-Tartaric acid salt, BMS 538305 tartarate, BMS-538305-06

  • CAS Number 841302-20-3 (free base)

  • Empirical Formula (Hill Notation) C17H26N2O2 · C4H6O6

  • Molecular Weight 440.49

Purchase

Properties

Related Categories B, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
Quality Level   100
assay   ≥98% (HPLC)
form   powder
application(s)   inhibition assay: suitable
color   white to beige
solubility   H2O: 20 mg/mL, clear
shipped in   ambient
storage temp.   room temp

Description

General description

BMS-538305 is a saxagliptin analogue, without a nitrile group.

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

BMS-538305, a saxagliptin analog, is a potent and selective dipeptidyl peptidase IV (DPP-IV) inhibitor.

Features and Benefits

BMS-538305 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Discover Bristol-Myers Squibb Compounds

We understand that in your research, the right tools are essential. Our commitment to providing researchers with the broadest selection of reliable, quality-controlled tools has resulted in a partner...
Keywords: Bioactive small molecules, Clinical, Genomics, Immunology, Neuroscience, Research areas

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?