• USA Home
  • BR81027S - Lymphocytic Choriomeningitis (LCM) Set

EMAIL THIS PAGE TO A FRIEND
BR81027S Sigma-Aldrich

Lymphocytic Choriomeningitis (LCM) Set

Cell culture extract

Synonym: LCM, LCMV

Purchase

Properties

form   liquid
species reactivity   mouse, rat
shipped in   dry ice
storage temp.   −70°C

Description

General description

LCM virus is a single stranded RNA virus that belongs to the Arenaviridae family. LCM virus is only transmitted by hamsters and mice; though infection of rats and other mammals is possible.

Application

This product has been tested in ELISA applications. When diluted sera is added to test wells coated with liquid antigen and control antigen, antibodies to hantavirus antigens will only bind in the antigen-coated wells. Labeled conjugate antibody will then allow for the detection of these antibodies through a chromogenic reaction with a substrate.

Other Notes

Cellular proteins from Mesocricetus auratus epithelial, BHK-21 cell line

Liquid antigen produced in vitro with coordinating cell line control antigen. The set is supplied in a 480 reaction format.

Kit component only

Description

Product #

Add to Cart

Lymphocytic Choriomeningitis (LCM) Liquid Antigen, Cell culture extract    
Cell Line Control Antigen for LCM, Cell culture extract    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?