EMAIL THIS PAGE TO A FRIEND
C0457 Sigma-Aldrich

Benzofuran-3(2H)-one

≥98% (HPLC)

Synonym: 3-Coumaranone

  • CAS Number 7169-34-8

  • Empirical Formula (Hill Notation) C8H6O2

  • Molecular Weight 134.13

  •  Beilstein/REAXYS Number 115296

  •  MDL number MFCD00051810

Purchase

Properties

assay   ≥98% (HPLC)
SMILES string   O=C1COc2ccccc12
InChI   1S/C8H6O2/c9-7-5-10-8-4-2-1-3-6(7)8/h1-4H,5H2
InChI key   MGKPCLNUSDGXGT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

12502 Benzofuran-3(2H)-one, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?