• USA Home
  • C0855 - Z-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrobromide

EMAIL THIS PAGE TO A FRIEND
C0855 Sigma-Aldrich

Z-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrobromide

≥85% (TLC), powder

Synonym: Z-GPR-AMC·HBr

  • Linear Formula C31H37N7O7·HBr

  • Molecular Weight 700.58

  •  MDL number MFCD03452878

Purchase

Description

Amino Acid Sequence

Z-Gly-Pro-Arg-AMC

Application

Fluorogenic substrate for granzyme A.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?