• USA Home
  • C1110AO-HH - DMT-dC(bz) Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
C1110AO-HH Sigma-Aldrich

DMT-dC(bz) Phosphoramidite

configured for ÄKTA® and OligoPilot®

Synonym: Cytidine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-dC(bz) amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-?bis(1-methylethyl)phosphoramidite], N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Purchase

Properties

Related Categories ABI 3900, ABI 39X, Chemical Synthesis, DNA Phosphoramidites, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents),
biological source   non-animal source
InChI Key   PGTNFMKLGRFZDX-SALLYJDFSA-N
type   for DNA synthesis
product line   Proligo Reagents
assay   ≥99% (31P-NMR)
  ≥99.0% (reversed phase HPLC)
form   powder or granules
packaging   pkg of 1 × 10 g (C111010-01)
  pkg of 1 × 5 g (C111005-01)
  pkg of 6 × 5 g (C111005-06)
application(s)   oligo synthesis: suitable
impurities   ≤0.3 wt. % water content (Karl Fischer)
color   white to off-white
λ   conforms (UV/VIS Identity)
compatibility   configured for ÄKTA® and OligoPilot®
nucleoside profile   base: cytidine
base protecting group: benzoyl
5' protecting group: DMT
deprotection: standard
storage temp.   2-8°C

Description

Legal Information

OligoPilot is a registered trademark of GE Healthcare

ÄKTA is a registered trademark of GE Healthcare

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?