• USA Home
  • C212080 - DMT-2′O-TBDMS-rC(tac) Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
C212080 Sigma-Aldrich

DMT-2′O-TBDMS-rC(tac) Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym: DMT-2′-O-TBDMS-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-?methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]

  • CAS Number 149989-66-2

  • Empirical Formula (Hill Notation) C57H76N5O10PSi

  • Molecular Weight 1050.30

  •  MDL number MFCD09842117

  •  PubChem Substance ID 329774935

  •  NACRES NA.51

Purchase

Properties

Related Categories 2′O-TBDMS RNA Phosphoramidites, Chemical Synthesis, DMT-2′O-TBDMS-rC(tac) Phosphoramidite, Expedite, Fast Deprotection 2′O-TBDMS RNA Phosphoramidites,
Quality Level   200
product line   Proligo Reagents
assay   ≥98% (31P-NMR)
  ≥98.0% (reversed phase HPLC)
application(s)   oligo synthesis: suitable
λ   conforms (UV/VIS Identity)
compatibility   configured for PerkinElmer
  configured for Polygen
nucleoside profile   base: cytidine
base protecting group: TAC
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast
storage temp.   −20°C
SMILES string   COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(=O)COc4ccc(cc4)C(C)(C)C)=NC3=O)(c5ccccc5)c6ccc(OC)cc6
InChI   1S/C57H76N5O10PSi/c1-39(2)62(40(3)4)73(69-36-18-34-58)71-51-48(37-68-57(42-19-16-15-17-20-42,43-23-27-45(65-11)28-24-43)44-25-29-46(66-12)30-26-44)70-53(52(51)72-74(13,14)56(8,9)10)61-35-33-49(60-54(61)64)59-50(63)38-67-47-31-21-41(22-32-47)55(5,6)7/h15-17,19-33,35,39-40,48,51-53H,18,36-38H2,1-14H3,(H,59,60,63,64)/t48-,51-,52-,53-,73?/m1/s1
InChI key   FAODNIJMAKFTFL-WSFKJFENSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?