• USA Home
  • C21213-HH - DMT-2′O-Methyl-rC(tac) Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
C21213-HH Sigma-Aldrich

DMT-2′O-Methyl-rC(tac) Phosphoramidite

configured for ABI

Synonym: DMT-2′-O-Me-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]

Purchase

Properties

Related Categories 2′O-Methyl RNA Phosphoramidites, ABI 3900, ABI 39X, Chemical Synthesis, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents),
InChI Key   JRJMGDLRRSEUHM-MVHRPHEDSA-N
product line   Proligo Reagents
assay   ≥98.0% (reversed phase HPLC)
  ≥99% (31P-NMR)
packaging   pkg of 1 × 0.5 g (C212131-01)
  pkg of 1 × 1 g (C212161-01)
application(s)   oligo synthesis: suitable
λ   conforms (UV/VIS Identity)
compatibility   configured for ABI
nucleoside profile   base: cytidine
base protecting group: TAC
2' protecting group: methyl
5' protecting group: DMT
deprotection: fast
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?