• USA Home
  • C2976 - Anti-Caspase 8 (GD-13) antibody produced in rabbit

C2976 Sigma

Anti-Caspase 8 (GD-13) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution



Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Caspases,
species reactivity   human
application(s)   microarray: suitable
  western blot: 1:1,000 using human Burkitt lymphoma Raji cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen (caspase 8a) mol wt 55 kDa
  antigen (caspase 8b) mol wt 54 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CASP8(841)
biological source   rabbit
conjugate   unconjugated



synthetic peptide of the human caspase 8a/MACHα1 precursor (amino acids 362-374 with N-terminal added lysine), conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.


The sequence used as the immunogen corresponds to the C-terminal region of the enzyme p18 subunit and is identical in other human caspase 8 isoforms (MACHα2, MACHα3, MACHβ) and has limited homology (approx. 50%) to mouse caspase 8. By immunoblotting, the antibody reacts with caspase 8a and caspase 8b.


Anti-Caspase 8 (GD-13) is suitable for microarray. It is also suitable for western blot at a concentration of 1:1,000 using human Burkitt lymphoma Raji cell extract.

Biochem/physiol Actions

Caspase 8 cleaves and activates CASP3, CASP4, CASP6, CASP7, CASP9 and CASP10. May participate in the GZMB apoptotic pathways. It also cleaves ADPRT. Caspase 8 hydrolyzes the small-molecule substrate, Ac-Asp-Glu-Val-Asp-|-AMC. It is also a target for cowpox virus CRMA death inhibitory protein. Anti-Caspase 8 (GD-13) recognizes human caspase 8a (55 kDa) and caspase 8b (54kDa) by immunoblotting. Staining of caspase 8 in immunoblotting is specifically inhibited with caspase 8 immunizing peptide (human, amino acids 362-374). Caspases play a crucial role in apoptosis in almost every cell type.

Price and Availability

Explore with Confidence

Expore Enzymes, Proteins & Cofactors

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?