• USA Home
  • C361010 - DMT-dC(bz)-CPG column

EMAIL THIS PAGE TO A FRIEND
C361010 Sigma-Aldrich

DMT-dC(bz)-CPG column

pore size 500 Å, 1 μmol, configured for PerkinElmer, configured for ABI

Purchase

Properties

Related Categories ABI 39X, Chemical Synthesis, Columns for Synthesizer, Controlled Pore Glass (CPG), DNA CPG 500A Integral Columns for ABI and Expedite,
type   for DNA synthesis
product line   Proligo Reagents
assay   ≥95.0% (Nucleoside Purity, reversed phase HPLC)
packaging   pkg of 1 × 100 columns
pore size   500 Å
capacity   ≥99 % (Coupling Efficiency)
λ   conforms (UV/VIS Identity)
compatibility   configured for ABI
  configured for PerkinElmer
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?