EMAIL THIS PAGE TO A FRIEND
C4146 Sigma-Aldrich

CM-Cellulose

preswollen, microgranular

Synonym: Carboxymethyl cellulose

Purchase

Properties

form   preswollen, microgranular
contains   77-81% moisture content
capacity   0.60-0.85 g/g, dry material capacity for proteins(lysozyme in 0.1M sodium acetate buffer, pH 5.0)
  0.90-1.15 meq/g, dry material small ion capacity
  1.05-1.45 g/g capacity for proteins(dry weight (0.01M sodium acetate buffer, pH 5.0))
InChI   1S/C6H12O6.C2H4O2/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4/h1,3-6,8-12H,2H2;1H3,(H,3,4)
InChI key   VJHCJDRQFCCTHL-UHFFFAOYSA-N

Description

Application

CM Cellulose products are used in protein chromatography, ion exchange chromatography and cation exchange media. CM Cellulose has been used in studies to inform the suppressed proliferation of hepatoma and leukemic cells and the inhibition of the human immunodeficiency virus type 1 (HIV-1).

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?