• USA Home
  • C423060 - DMT-dC(ac)-CPG Column

EMAIL THIS PAGE TO A FRIEND
C423060 Sigma-Aldrich

DMT-dC(ac)-CPG Column

0.2 μmol, pore size 1000 Å, configured for ABI, configured for PerkinElmer

Purchase

Properties

Related Categories ABI 39X, Chemical Synthesis, DNA Column, Expedite, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents),
type   for DNA synthesis
product line   Proligo Reagents
  Proligo Reagents
assay   ≥95.0% (Nucleoside Purity, reversed phase HPLC)
pore size   1000 Å
capacity   ≥99.0 % (Coupling Efficiency)
λ   conforms (UV/VIS Identity)
compatibility   configured for ABI
  configured for PerkinElmer
storage temp.   −20°C
  −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?