• USA Home
  • C4387 - Collagen from bovine achilles tendon

EMAIL THIS PAGE TO A FRIEND
C4387 Sigma-Aldrich

Collagen from bovine achilles tendon

powder, suitable for substrate for collagenase

Purchase

Properties

Related Categories Biochemicals and Reagents, Collagen, Proteins and Derivatives, Structural Proteins
biological source   bovine Achilles tendon
form   powder
suitability   suitable for substrate for collagenase
storage temp.   2-8°C
Gene Information   bovine ... COL1A1(282187)

Description

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Collagen terminology using the Bornstein and Traub designation originates from the reference; Bornstein, P. and Traub, W. The Proteins, (1979) 4, 411-605

Application

Bovine achilles tendon collagen was used in a study of plant extracts as inhibitors of MMP-collagenases, the putative active agents in the breakdown of cartilage in osteoarthritis and rheumatoid arthritis.

Collagen from bovine Achilles tendon is a naturally occurring protein in the form of elongated fibrils. It may be used in studies of the fibrocartilaginous zone, which the collagen must first pass through before inserting into the calcaneus. It may also be used in studies of growth factor effects on collagen content and cross-linking during Achilles tendon healing.

Preparation Note

Prepared by the method of Einbinder, J. and Schubert, M., J. Biol. Chem., 188, 335 (1951).

Reconstitution

This product is an insoluble collagen preparation. It is insoluble in water, aqueous buffers, dilute acid, and organic solvents. For use as a substrate in collagenase assays, this collagen can be prepared as a suspension in 50 mM TES buffer, pH 7.4 with 0.36 mM calcium chloride.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
How should Product No. C4387 be reconstituted?
This product is an insoluble collagen preparation. It is insoluble in water, aqueous buffers, dilute acid, and organic solvents. A suspension can be prepared in 50 mM TES buffer, pH 7.4 with 0.36 mM calcium chloride, but not a clear solution.
What application can Product No. C4387 be used in?
This product is suitable for use as a substrate for collagenase. It is not suitable for use as an attachment factor in coating glassware.
What "type" of collagen does Product No. C4387 consist of?
This collagen is classified as Bornstein and Traub type I. It is not to be confused with Sigma's catalog type which is an organizational placeholder.
What is the structure of the protein Collagen from bovine, ProductC4387?
The amino acid sequence of the primary structure is mainly a repeating motif with glycine in every third position, and a proline or 4-hydroxyproline frequently preceding the glycine residue.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?