EMAIL THIS PAGE TO A FRIEND
C4393 Sigma-Aldrich

Cholesteryl iodide

~98%

Purchase

Properties

assay   ~98%
storage temp.   −20°C
SMILES string   CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](I)CC[C@]4(C)[C@H]3CC[C@]12C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?