EMAIL THIS PAGE TO A FRIEND
C4404 Supelco

Clonazepam solution

drug standard, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Benzodiazepin Analoga Structure, Benzodiazepines,
grade   drug standard
assay   ≥98%
drug control   Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C
SMILES string   [O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3Cl)c2c1
InChI   1S/C15H10ClN3O3/c16-12-4-2-1-3-10(12)15-11-7-9(19(21)22)5-6-13(11)18-14(20)8-17-15/h1-7H,8H2,(H,18,20)
InChI key   DGBIGWXXNGSACT-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Articles

HPLC Analysis of Antianxiety Drugs on Ascentis® Express C18 vs. Standard C18, Demonstrating High Throughput on Ascentis Express

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antianxiety Drugs on Ascentis® Express C18 vs. Standard C18, Demonstrating High Throughput on Ascentis Express
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Antianxiety Drugs on Ascentis® Phenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antianxiety Drugs on Ascentis® Phenyl
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Anticonvulsant Drugs on Discovery® RP-AmideC16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Anticonvulsant Drugs on Discovery® RP-AmideC16
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Anticonvulsants on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Anticonvulsants on Discovery® C18
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Anticonvulsants on Discovery® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Anticonvulsants on Discovery® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® C8
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Mass spectrometry, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® Express C18, Acetonitrile Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® Express C18, Acetonitrile Mobile Phase
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl with Acetonitrile Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl with Acetonitrile Mobile Phase
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl, Methanol Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl, Methanol Mobile Phase
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

RP-HPLC Analysis of Benzodiazepines on Ascentis® Express Biphenyl

From our library of Articles, Sigma-Aldrich presents RP-HPLC Analysis of Benzodiazepines on Ascentis® Express Biphenyl
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C1277 Clonazepam, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?