EMAIL THIS PAGE TO A FRIEND
C5931 Sigma-Aldrich

β-Cyclocitral

≥70% (GC)

Synonym: 2,6,6-Trimethyl-1-cyclohexene-1-carboxaldehyde

Purchase

Properties

assay   ≥70% (GC)
refractive index   n20/D 1.497 (lit.)
bp   62-63 °C/3 mmHg (lit.)
density   0.943 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   [H]C(=O)C1=C(C)CCCC1(C)C
InChI   1S/C10H16O/c1-8-5-4-6-10(2,3)9(8)7-11/h7H,4-6H2,1-3H3
InChI key   MOQGCGNUWBPGTQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?