• USA Home
  • C5979 - Anti-CD30 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
C5979 Sigma-Aldrich

Anti-CD30 antibody produced in goat

affinity isolated antibody, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Analytical Cytology, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology,
conjugate   unconjugated
clone   polyclonal
biological source   goat
application(s)   indirect ELISA: 0.5-1.0 μg/mL
  western blot: 0.1-0.2 μg/mL
species reactivity   human
form   lyophilized powder
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   P28908
Gene Information   human ... TNFRSF8(943)

Description

General description

CD30 is a type I transmembrane glycoprotein of the TNF receptor superfamily. CD30 was originally identified as a cell surface antigen of Hodgkin’s and Reed-Sternberg cells using monoclonal antibody Ki-1.

Specificity

Shows <1% cross-reactivity with recombinant mouse CD30.

Immunogen

recombinant human CD30, extracellular domain, expressed in Sf21 cells.

Application

Anti-CD30 antibody produced in goat is suitable for indirect ELISA at a concentration of 0.5-1.0μg/mL and western blotting at a concentration of 0.1-0.2μg/mL.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Reconstitution

Reconstitute with 1 mL sterile PBS to yield 0.1 mg/ml solution

Biochem/physiol Actions

CD30 has a critical role in the pathophysiology of Hodgkin’s disease and other CD30+ lymphomas. CD30 acts as a costimulatory molecule in thymic negative selection. CD30 interacts with tumor necrosis factor receptor-associated factor (TRAF)1 and TRAF2, and induces cell death in T cell hybridomas. Similarly, CD30 also interacts with TRAF2 and TRAF5 to regulate nuclear factor κB (NFκB) activation. CD30 is also crucial for human embryonic stem cells survival and proliferation via NFκB activation.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?