• USA Home
  • C5983 - Collagen from human lung

EMAIL THIS PAGE TO A FRIEND
C5983 Sigma-Aldrich

Collagen from human lung

Bornstein and Traub Type I, powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Collagen, Proteins and Derivatives, Structural Proteins
biological source   human lung
type   Type I
form   powder
application(s)   cell culture | stem cell: suitable
storage temp.   2-8°C
Gene Information   human ... COL1A1(1277), COL1A2(1278), COL27A1(85301), COL3A1(1281), COL5A1(1289)

Description

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Application

Collagen Type I has been used as a scaffold for the growth in vitro of stem cells in a wide variety of biomaterial engineering studies.

Collagen-type 1 may be used in research of Idiopathic pulmonary fibrosis (IPF). Robust expression of collagen-type 1 is one distinctive feature of IPF. Additionally, collagen-type 1 has been used in studies on the effect of endoplasmic reticulum (ER) stress from IPF on myofibroblastic differentiation of lung fibroblasts. Collage-type 1 soluble in acidic solution produces three dimensional scaffolding useful in bioengineering and cell culture applications where biomaterials are needed to replace native collagen extracellular matrices.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?