EMAIL THIS PAGE TO A FRIEND
C6128 Sigma-Aldrich

6-Chloropurine

crystalline

  • CAS Number 87-42-3

  • Empirical Formula (Hill Notation) C5H3ClN4

  • Molecular Weight 154.56

  •  Beilstein/REAXYS Number 5774

  •  EC Number 201-745-6

  •  MDL number MFCD00075825

  •  PubChem Substance ID 329775114

Purchase

Properties

form   crystalline
color   white to faint yellow
mp   >300 °C (dec.) (lit.)
storage temp.   2-8°C
SMILES string   Clc1ncnc2[nH]cnc12
InChI   1S/C5H3ClN4/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H,7,8,9,10)
InChI key   ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UO7520000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

511617 6-Chloropurine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?