• USA Home
  • C6298 - 6-Chloropurine 2′-deoxyriboside

EMAIL THIS PAGE TO A FRIEND
C6298 Sigma-Aldrich

6-Chloropurine 2′-deoxyriboside

≥95%

  • CAS Number 4594-45-0

  • Empirical Formula (Hill Notation) C10H11ClN4O3

  • Molecular Weight 270.67

  •  MDL number MFCD00083282

Purchase

Properties

assay   ≥95%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?