EMAIL THIS PAGE TO A FRIEND
C7198 Sigma

Clozapine

drug standard, 1.0 mg/mL in methanol

Synonym: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine

  • CAS Number 5786-21-0

  • Empirical Formula (Hill Notation) C18H19ClN4

  • Molecular Weight 326.82

  •  MDL number MFCD00153785

  •  PubChem Substance ID 329775141

Purchase

Properties

grade   drug standard
InChI Key   QZUDBNBUXVUHMW-UHFFFAOYSA-N
assay   ≥98%
concentration   1.0 mg/mL in methanol
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?