• USA Home
  • C7349 - Anti-Caspase 2 antibody produced in rabbit

C7349 Sigma

Anti-Caspase 2 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Ich-1L



Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Caspases,
species reactivity   mouse, rat, human, chicken
application(s)   microarray: suitable
  western blot: 1:200 using Jurkat human T-cell leukemia cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 48 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CASP2(835)
mouse ... Casp2(12366)
rat ... Casp2(64314)
biological source   rabbit
conjugate   unconjugated



synthetic peptide corresponding to the C-terminal of human procaspase 2 (amino acids 420-435 with N-terminal added lysine) conjugated to KLH. This sequence is highly conserved in rat, mouse, and chicken.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.


The antibody reacts specifically with caspase 2 (Ich-1L).

Price and Availability

Explore with Confidence

Expore Enzymes, Proteins & Cofactors

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?