• USA Home
  • C7696 - Cyclophilin A from calf thymus

EMAIL THIS PAGE TO A FRIEND
C7696 Sigma-Aldrich

Cyclophilin A from calf thymus

powder

Synonym: CyP

Purchase

Description

Packaging

0.1 mg in poly bottle

Biochem/physiol Actions

Cyclophilins are peptidyl prolyl isomerases that catalyze the cis-trans isomerization of X-Pro peptide bonds. They are highly-conserved cytoplasmic enzymes that accelerate protein folding and facilitate HIV infectivity. Cyclosporin A binds to cyclophilin and inhibits its activity. The cyclosporin A-cyclophilin complex binds to calcineurin and inhibits T-cell activation. The structure of human, recombinant cyclophilin is given by Holzman, et al.

Quality

Highly purified, lyophilized powder, balance primarily NaCl, HEPES, DTT, PMSF, sodium azide and stabilizer.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?