• USA Home
  • C7919 - 4-Chlorotestosterone 17-acetate

EMAIL THIS PAGE TO A FRIEND
C7919 Sigma

4-Chlorotestosterone 17-acetate

≥95%

Synonym: 17β-Acetoxy-4-chloroandrost-4-en-3-one, 4-Chloro-17β-hydroxy-4-androsten-3-one 17-acetate, 4-Chlorotestosterone 17-acetate, Clostebol acetate

Purchase

Properties

assay   ≥95%
drug control   USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
storage temp.   2-8°C
Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
UN 2252 3 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?