• USA Home
  • C8227 - Anti-CXCR4 (Fusin), N-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C8227 Sigma-Aldrich

Anti-CXCR4 (Fusin), N-Terminal antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Proteins/Bioactives/Markers/Receptors for Stem Cell Biology, Antibodies for Cell Biology, Antibodies for Hematopoietic Stem Cells,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 1-2 μg/mL using human HeLa cell extract.
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
UniProt accession no.   P61073
Gene Information   human ... CXCR4(7852)

Description

General description

Chemokine receptor CXCR-4 also refers as fusin or LESTR is a protein that belongs to G-protein coupled receptor 1 family. It is an essential cofactor for T cell-tropic HIV viruses into CD4+ cells. N-Terminal domain and second extracellular loop of the protein act as a HIV binding sites. Anti-CXCR4 (Fusin), N-Terminal antibody can be used to block HIV-1 and HIV-2 fusion and infection of human target cells. It can also be used in immunoprecipitation. Rabbit anti- CXCR4 (Fusin), N-Terminal antibody reacts specifically with human CXCR4.

Immunogen

synthetic peptide corresponding to amino acids 1-14 of the N-terminal of human CXCR4.

Application

Anti-CXCR4 (Fusin), N-Terminal antibody can be used in immunoblotting and immunocytochemistry.

Physical form

Solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Other Notes

The antigen (receptor) is also termed fusin, LESTER and HUMSTR.

Biochem/physiol Actions

CXCR4 is a cell surface receptor of the chemokine CXCL12 (SDF-1) and is involved in metastasis, hematopoiesis and immune responses. It acts with CD4 to facilitate the entry of HIV into target cells. CXCR4 has important roles in tumor initiation, proliferation, invasion and metastasis of breast cancer cells.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Cancer Stem Cells: New Targets for Cancer Therapy

Interested in receiving more content like this? Join over 8,000 of your peers who have already signed up for the Cell Informer newsletter.
Keywords: Cancer, Cell attachment, Cell culture, Cell proliferation, Gastrointestinal, Gene expression, Transcription, transformation

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?