• USA Home
  • C8448 - Chartreusin from Streptomyces chartreusis

EMAIL THIS PAGE TO A FRIEND
C8448 Sigma-Aldrich

Chartreusin from Streptomyces chartreusis

Synonym: Lambdamycin

  • CAS Number 6377-18-0

  • Empirical Formula (Hill Notation) C32H32O14

  • Molecular Weight 640.59

  •  MDL number MFCD00467138

Purchase

Properties

storage temp.   2-8°C
SMILES string   CO[C@H]1[C@@H](O)[C@@H](C)O[C@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](C)O[C@H]2Oc3cccc4c(O)c5C(=O)Oc6ccc(C)c7C(=O)Oc(c5-c67)c34)[C@@H]1O

Description

Biochem/physiol Actions

Antitumor antibiotic which induces single-strand scission in DNA in the presence of reducing agents.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FL7350000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?