EMAIL THIS PAGE TO A FRIEND
Sigma

UM-UC-11

08090507

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   human bladder
growth mode   Adherent
karyotype   Not specified
morphology   Epithelial
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
relevant disease(s)   cancer
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Depositor and originator: Dr H B Grossman Department of Urology, University of Texas, M.D. Anderson Cancer Centre, 1515 Holcombe Boulevard, Unit 1373, Houston, Texas, 77030 USA

Cell Line Origin

Human transitional cell carcinoma of the bladder

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 10
D13S317: 11,14
D16S539: 11
D5S818: 12
D7S820: 8,12
THO1: 6,9
TPOX: 9
vWA: 14,17

Culture Medium

EMEM (EBSS) + 2 mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37 °C.

Cell Line Description

UM-UC-11 was derived from a transitional cell carcinoma of the bladder. UM-UC-11 failed to produce tumours in athymic nude mice after 90 days. Susceptibility to adenoviral mediated gene transduction and expression of p53, p16, retinoblastoma protein and coxsackie adenovirus receptor were studied.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis


Cell Culture Guide

ECACC Laboratory Handbook
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?