• USA Home
  • Trex1 KO Cell line from mouse

EMAIL THIS PAGE TO A FRIEND
Sigma

Trex1 KO Cell line from mouse

12021509

Synonym: Trex1-KO

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   mouse embryo
growth mode   adherent
morphology   fibroblast
shipped in   dry ice
storage temp.   −196°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Cell Culture Guide

ECACC Laboratory Handbook
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?