EMAIL THIS PAGE TO A FRIEND
Sigma

WRC

88031148

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   rat unknown/unspecified
growth mode   Adherent
karyotype   2n = 42
morphology   Epithelial
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
relevant disease(s)   cancer
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Depositor and originator: Dr E I Levene, Department of Pathology, University of Cambridge

Application

Tumourigenicity and virus studies: HSV, PRV and vaccinia viruses

Cell Line Origin

Rat Walker carcinoma

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

Derived from the parent line LLC-WRC 256, which was established in 1954. The cells are tumorigenic in suckling rats and support the growth of HSV, PRV and Vaccinia viruses.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis


Cell Culture Guide

ECACC Laboratory Handbook
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?