EMAIL THIS PAGE TO A FRIEND
Sigma

SV-T2

90030801

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   mouse embryo
growth mode   Adherent
karyotype   2n = 40, modal No 41
morphology   Fibroblast
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Cell Line Origin

Mouse BALB/3T3 clone A31

Cell Line Description

Derived by SV40 transformation of the line BALB/3T3 clone A31. The cells exhibit a high saturation density, lack contact inhibition and form tumours in appropriate hosts.

Application

Transformation, tumourigenicity and virus studies: SV40

Culture Medium

DMEM + 2mM Glutamine + 10% New Born Calf Serum (NBCS) or (5% NBCS + 5% Foetal Bovine Serum (FBS)).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1x1,000 to 3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Cell Culture Guide

ECACC Laboratory Handbook
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?