EMAIL THIS PAGE TO A FRIEND

C211

90112604

Purchase

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   human skin
growth mode   Adherent
karyotype   2n = 46
morphology   Fibroblast
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Cell Line Origin

Human Caucasian skin, Cri du Chat syndrome

Cell Line Description

Derived from a biopsy of an 11 year old Caucasian male with Cri du Chat syndrome. Cells have a limited lifespan of approximately 35 passages. Autoradiographic studies demonstrated that a distinction could be made between chromosome 4 and 5 in the Cri du chat syndrome by alternative DNA replication pattern and length of long and short arms.

Application

Genetic studies

Culture Medium

McCoy′s 5a + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?