EMAIL THIS PAGE TO A FRIEND

GEEP

92062401

Purchase

Properties

Related Categories Cell Culture, D-I, ECACC Cell Lines More...
biological source   ovine hybrid
growth mode   Adherent
karyotype   2n = 57
morphology   Fibroblast
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Cell Line Origin

Goat x Sheep hybrid, skin.

Cell Line Description

Skin fibroblast derived from a 6 year old female goat x sheep interspecies hybrid (Capra Lircus x Ovis Aries).

Culture Medium

EMEM α (nucleotide enriched) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Pipette Controller from Brand
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?