EMAIL THIS PAGE TO A FRIEND
Sigma

STE-137

95122020

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, S-Z More...
biological source   fish embryo
growth mode   Adherent
karyotype   Modal no. 58
morphology   Epithelial
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Depositor and originator: Dr J L Fryer, Department of Microbiology, Oregon State University, Corvallis

Application

Isolation of viruses from clinical samples, virus propagation

Cell Line Origin

Fish steelhead trout embryo

Culture Medium

MEM (EBSS) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 18°C. Population doubling can take up to 72 hours. Fish cell lines detach easily during transit if the culture is very young. After resuscitation split cells 1:3 and culture for at least one week with 1-2 media changes before shipping.

Cell Line Description

STE-137 was initiated in 1963 from pooled steelhead embryos (Oncorynchus mykiss). This heteroploid cell line, also known as steelhead embryo-137, replicates and detects common salmonid viruses (infectious pancreatic necrotic virus (IPNV), infectious hematopoietic necrosis virus (IHNV) and viral hemorrhagic septicaemia virus (VHSV)). The growth temperature ranges from 15-21°C with a routine propagation at 18°C.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis


Cell Culture Guide

ECACC Laboratory Handbook
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?