• USA Home
  • CLS3706 - Corning® 384 well microplate

EMAIL THIS PAGE TO A FRIEND
CLS3706

Corning® 384 well microplate

white polystyrene, flat bottom, non-sterile, lid: no, case of 100

Purchase

Properties

Related Categories 384 Well Plates, Corning, Corning 384-Well Assay Plates, Corning 384-Well Microplates, Corning 384-Well Multiwell Plates,
material   flat bottom
  white polystyrene
sterility   non-sterile
feature   lid: no
packaging   case of 100
mfr. no.   Corning, 3706
maximum volume   112 μL
size   384 wells
working volume   80 μL
color   white
matrix active group   Non-treated surface treatment
binding type   non-treated surface

Description

General description

For cell-based luminescence and radiometric and some fluorescence assays

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Corning Microplate Selection Product Line

From our library of Related Content, Sigma-Aldrich presents Corning Microplate Selection Product Line
Keywords: Cell culture

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?