• USA Home
  • CLS3712BC - Corning® 384 well microplate, low volume

EMAIL THIS PAGE TO A FRIEND
CLS3712BC Sigma

Corning® 384 well microplate, low volume

matrix active group TC-Treated, black polystyrene, flat bottom, sterile, (with generic barcode), lid, pack of 10

Synonym: 384 multiwell plate, 384 well barcode plate, 384 well microplate, 384 well microtiter plate, 384 well plate with generic barcode, 384 well plate

Purchase

Properties

Related Categories 384 Well Plates, Corning, Corning 384-Well Multiwell Plates, Corning Barcoded Microplates, Corning Microplates,
material   black plate
  black polystyrene
  flat bottom
sterility   sterile
feature   lid
  (with generic barcode)
packaging   pack of 10
mfr. no.   Corning, 3712BC
matrix active group   TC-Treated
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?