• USA Home
  • CLS6530 - Corning® PCR tubes

EMAIL THIS PAGE TO A FRIEND
CLS6530

Corning® PCR tubes

0.5 mL PCR tube, polypropylene, flat cap, non-sterile, clear, 1000/cs

Synonym: 0.5 mL PCR tubes, PCR tubes

Purchase

Properties

Related Categories Corning, Corning PCR Tubes, Corning PCR tubes, Corning Tubes, Labware,
material   conical bottom tube
  flat cap
  polypropylene
sterility   non-sterile
feature   RCF 10,000 × g
  RNase and DNase free
  autoclavable at 121 °C
  Bottom style: conical
packaging   case of 1000
  pack of 250
mfr. no.   Corning, 6530
capacity   0.5 mL
tube O.D.   0.390 in.
color   clear
  clear
fitting   cap (push fit)
suitability   suitable for (PCR applications)

Description

General description

Corning® PCR Tubes

• Made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer
• Nonsterile and certified DNase- and RNase-free
• Autoclavable at 121°C and will withstand centrifugation to 10,000 x g

Legal Information

Corning is a registered trademark of Corning, Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Metagenomics and Quantitative PCR

The highly variable regions within the 16S ribosomal RNA (rRNA) gene sequences are being exploited with PCR and metagenomic sequencing to characterize the microbial strains within the gastrointestina...
Chloe McClanahan
Biofiles Edition 6.2
Keywords: Gastrointestinal, Nucleic acid hybridization, PAGE, Polymerase chain reaction, Polymerase chain reaction - quantitative, Probiotics, Sequencing

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?