• USA Home
  • D0438 - Monoclonal Anti-Dynamin antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
D0438 Sigma-Aldrich

Monoclonal Anti-Dynamin antibody produced in mouse

clone D5, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

clone   D5, monoclonal
application(s)   immunoprecipitation (IP): suitable
  indirect immunofluorescence: suitable
  western blot: 0.5-3 μg/mL using rat or mouse brain extract.
species reactivity   human, mouse, rat
mol wt   antigen 98 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C
antibody form   purified immunoglobulin
isotype   IgG1

Description

Specificity

By immunoblotting, the antibody specifically recognizes native dynamin.

Immunogen

recombinant human dynamin (amino acids 1-750)

Physical form

Solution in 0.05 M sodium phosphate buffer containing 0.2% gelatin and 0.1% sodium azide

Preparation Note

The antibody is purified using protein A and G.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?