• USA Home
  • D125 - Desipramine hydrochloride

EMAIL THIS PAGE TO A FRIEND
D125 Sigma-Aldrich

Desipramine hydrochloride

  • CAS Number 58-28-6

  • Empirical Formula (Hill Notation) C18H22N2 · HCl

  • Molecular Weight 302.84

  •  EC Number 200-373-1

  •  MDL number MFCD00058108

Purchase

Properties

SMILES string   Cl[H].CNCCCN1c2ccccc2CCc3ccccc13
InChI   1S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19H,6,11-14H2,1H3;1H
InChI key   XAEWZDYWZHIUCT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
HO0525000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D3900 Desipramine hydrochloride, ≥98% (TLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?