EMAIL THIS PAGE TO A FRIEND
D1398 Sigma-Aldrich

Dynorphin B porcine

≥95% (HPLC)

Synonym: Prodynorphin 228-240, Rimorphin

  • CAS Number 85006-82-2

  • Empirical Formula (Hill Notation) C74H115N21O17

  • Molecular Weight 1570.84

  •  MDL number MFCD00076372

  •  PubChem Substance ID 329798691

Purchase

Properties

assay   ≥95% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@@H](C)O)C(O)=O
InChI   1S/C74H115N21O17/c1-40(2)34-53(91-68(107)54(36-44-18-10-8-11-19-44)86-58(100)39-84-57(99)38-85-62(101)48(76)35-46-25-27-47(97)28-26-46)67(106)89-51(24-17-33-83-74(80)81)63(102)87-50(23-16-32-82-73(78)79)64(103)90-52(29-30-56(77)98)65(104)92-55(37-45-20-12-9-13-21-45)69(108)88-49(22-14-15-31-75)66(105)93-59(41(3)4)70(109)94-60(42(5)6)71(110)95-61(43(7)96)72(111)112/h8-13,18-21,25-28,40-43,48-55,59-61,96-97H,14-17,22-24,29-39,75-76H2,1-7H3,(H2,77,98)(H,84,99)(H,85,101)(H,86,100)(H,87,102)(H,88,108)(H,89,106)(H,90,103)(H,91,107)(H,92,104)(H,93,105)(H,94,109)(H,95,110)(H,111,112)(H4,78,79,82)(H4,80,81,83)/t43-,48+,49+,50+,51+,52+,53+,54+,55+,59+,60+,61+/m1/s1
InChI key   AGTSSZRZBSNTGQ-ITZCFHCWSA-N
Gene Information   pig ... PDYN(445529)

Description

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Gln-Phe-Lys-Val-Val-Thr

Biochem/physiol Actions

Endogenous κ1b opioid receptor agonist.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?