• USA Home
  • D1803 - (S)-N-(3,4-Dimethoxybenzyl)-α-phenylethylamine

EMAIL THIS PAGE TO A FRIEND
D1803 SAFC

(S)-N-(3,4-Dimethoxybenzyl)-α-phenylethylamine

≥99% (GC)

Purchase

Properties

assay   ≥99% (GC)

Description

Application

The lithium amide of this chiral base participates in diastereoselective Michael addition reactions, yielding β-amino acids as esters.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?