• USA Home
  • D217 - Anti-Dopamine-β-Hydroxylase (N-Terminal) antibody produced in sheep

D217 Sigma

Anti-Dopamine-β-Hydroxylase (N-Terminal) antibody produced in sheep

affinity isolated antibody, buffered aqueous glycerol solution



Related Categories Alphabetical Index, Antibodies, Antibodies for Neurobiology, Antibodies to Neural Enzymes, DM-DP,
species reactivity   sheep, canine, bovine, human, rat, rabbit
application(s)   western blot: 1:250
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... DBH(1621)
rat ... Dbh(25699)
biological source   sheep
conjugate   unconjugated



synthetic peptide from the N-terminal region of human dopamine-β-hydroxylase.

Physical form

Solution in 10% glycerol, 10 mM HEPES, pH 7.2, 20 mM Tris-HCl (pH 7.6), 1 mg/ml BSA, 0.1% PVP-40, 0.05% Tween 20, and 15 mM sodium azide.


By immunoblotting, the antibody reacts specifically with a single (or double) band at 70-75 kDa in samples of SDS-solubilized, DTT-reduced human adrenal medulla. No cross-reactivity is observed with monkey adrenal medulla.


Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability

Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?