• USA Home
  • D2545 - Desiccator safety cages
EMAIL THIS PAGE TO A FRIEND
D2545 Sigma
Desiccator safety cages
For 4 in. (10 cm) I.D. desiccator
Purchase
Properties
availability   not available in EU
Description
General description
Protects both the operator and the desiccator, these cages are constructed of 3/8 in. square-mesh wire coated with polyethylene. Specifically designed for use with the desiccators shown above.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?