• USA Home
  • D2647 - Dimethylamine solution

EMAIL THIS PAGE TO A FRIEND
D2647 Sigma-Aldrich

Dimethylamine solution

40% in H2O

  • CAS Number 124-40-3

  • Linear Formula (CH3)2NH

  • Molecular Weight 45.08

  •  Beilstein/REAXYS Number 605257

  •  MDL number MFCD00008288

Purchase

Properties

InChI Key   ROSDSFDQCJNGOL-UHFFFAOYSA-N
concentration   40% in H2O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1160 - class 3 - PG 2 - Dimethylamine solution
WGK Germany 
2
Flash Point(F) 
60.8 °F
Flash Point(C) 
16 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

426458 Dimethylamine solution, 40 wt. % in H2O

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?