• USA Home
  • D4319 - Anti-DYRK1B (516-530) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
D4319 Sigma-Aldrich

Anti-DYRK1B (516-530) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B isoform a (Homo sapiens), Anti-MIRK

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Epigenetics and Nuclear Signaling, Antibodies for Kinase/Phosphatase Biology,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 1:500-1:2,000
species reactivity   human
mol wt   antigen ~69 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
UniProt accession no.   Q9Y463
Gene Information   human ... DYRK1B(9149)

Description

General description

This gene encodes a member of a family of nuclear-localized protein kinases. It is expressed at low levels in most normal tissues but at elevated levels in many tumor cell lines and in normal skeletal muscle.

Immunogen

synthetic peptide corresponding to amino acids 516-530 of human DYRK1B.

Application

Anti-DYRK1B antibody produced in rabbit is suitable for western blot analysis at a working dilution of 1:500-1:2000. Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Biochem/physiol Actions

DYRK1B encodes a protein that is a dual-specificity kinase with both serine/ threonine and tyrosine kinase activities. It enhances the transcriptional activity of TCF1/HNF1A and FOXO1. It inhibits epithelial cell migration through opposing the action of the Met signaling cascade adaptor protein RanBPM (Ran-binding protein M). It is a mitogen-activated protein kinase substrate that mediates colon carcinoma cell survival in mitogen-poor environments.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?